• USA Home
  • Search Results

  • ampicillinsodiumsalt371396952311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ampicillin sodium salt

3 Products
Synonym:   D-(−)-α-Aminobenzylpenicillin sodium salt
Image of Ampicillin sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C16H18N3NaO4S
Molecular Weight:
 371.39
 Beilstein/REAXYS Number:
 4119211
 EC Number:
 MDL number:
 NACRES:
 NA.76
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Mode of Action
  Application(s)
  Solubility
  Product Line
  Color
  Physical Form
Show More