• USA Home
  • Search Results

  • ampicillintrihydrate40345717748211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ampicillin trihydrate

6 Products
Synonym:   D-(−)-α-Aminobenzylpenicillin
Image of Ampicillin trihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C16H19N3O4S · 3H2O
Molecular Weight:
 403.45
 Beilstein/REAXYS Number:
 5399534
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Mode of Action
  pK-Values
  Anal. Standard Form
  Manufacturer designation
  Activity Spectrum
  Physical Form
  Application(s)
Show More