• USA Home
  • Search Results

  • amylalcohol88157141011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Amyl alcohol

1 Products
Synonym:   1-Pentanol, n-Amyl alcohol, Pentyl alcohol
Image of Amyl alcohol
CAS Number:
Linear Formula:
 CH3(CH2)4OH
Molecular Weight:
 88.15
 Beilstein/REAXYS Number:
 1730975
 Council of Europe no.:
 514
 EC Number:
 FEMA Number:
 2056
 Flavis number:
 2.040
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
Show More

Product #
Description
W205605 Amyl alcohol, ≥99%, FG,