• USA Home
  • Search Results

  • amyloglucosidasefromaspergillusniger12345903208011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Amyloglucosidase from Aspergillus niger

6 Products
Synonym:   1,4-α-D-Glucan glucohydrolase, Exo-1,4-α-glucosidase, Glucoamylase
Image of Amyloglucosidase from <I>Aspergillus niger</I>
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.2.1.3  ( BRENDA | IUBMB )
 MDL number:
 NACRES:
 NA.54
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Molecular weight
Show More