• USA Home
  • Search Results

  • anhydrouslactose342306342311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Anhydrous lactose

1 Products
Synonym:   Lactose
Image of Anhydrous lactose
CAS Number:
Empirical Formula (Hill Notation):
 C12H22O11
Molecular Weight:
 342.30
 Beilstein/REAXYS Number:
 93796
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More