• USA Home
  • Search Results

  • anilinehydrochloride1295914204111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aniline hydrochloride

2 Products
Image of Aniline hydrochloride
CAS Number:
Linear Formula:
 C6H5NH2 · HCl
Molecular Weight:
 129.59
 Beilstein/REAXYS Number:
 3593823
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More