• USA Home
  • Search Results

  • anisylformate1661712291811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Anisyl formate

1 Products
Synonym:   4-Methoxybenzyl formate
Image of Anisyl formate
CAS Number:
Linear Formula:
 HCO2CH2C6H4OCH3
Molecular Weight:
 166.17
 Council of Europe no.:
 354
 EC Number:
 FEMA Number:
 2101
 Flavis number:
 9.087
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Special Grade
Show More