• USA Home
  • Search Results

  • anthraquinone208218465111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Anthraquinone

5 Products
Image of Anthraquinone
CAS Number:
Empirical Formula (Hill Notation):
 C14H8O2
Molecular Weight:
 208.21
 Beilstein/REAXYS Number:
 390030
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Physical Form
  Manufacturer designation
  Special Grade
  Product Line
  Application(s)
Show More