• USA Home
  • Search Results

  • anthrarufin2402111712411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Anthrarufin

1 Products
Synonym:   1,5-Dihydroxyanthraquinone
Image of Anthrarufin
CAS Number:
Empirical Formula (Hill Notation):
 C14H8O4
Molecular Weight:
 240.21
 Beilstein/REAXYS Number:
 1881718
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More