• USA Home
  • Search Results

  • antimonyiiioxide29152130964411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Antimony(III) oxide

6 Products
Synonym:   Antimony(III) oxide, di-Antimony trioxide
Image of Antimony(III) oxide
CAS Number:
Empirical Formula (Hill Notation):
 O3Sb2
Molecular Weight:
 291.52
 EC Index Number
 215-175-0
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Vapor pressure
  Solubility
  Potency
  Particle Dimension
  Melting Point
  Density
  Bulk Density
  Physical Form
Show More