• USA Home
  • Search Results

  • antimonyiiisulfide33972134504611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Antimony(III) sulfide

2 Products
Image of Antimony(III) sulfide
CAS Number:
Linear Formula:
 Sb2S3
Molecular Weight:
 339.72
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
 eCl@ss:
 38080208
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More