• USA Home
  • Search Results

  • antimonyvoxide32352131460911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Antimony(V) oxide

1 Products
Synonym:   Antimonic oxide, Antimony pentoxide, Diantimony pentaoxide, Diantimony pentoxide
Image of Antimony(V) oxide
CAS Number:
Linear Formula:
 Sb2O5
Molecular Weight:
 323.52
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More