• USA Home
  • Search Results

  • antu202288688411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

ANTU

1 Products
Synonym:   N-(1-Naphthyl)thiourea
Image of ANTU
CAS Number:
Empirical Formula (Hill Notation):
 C11H10N2S
Molecular Weight:
 202.28
 Beilstein/REAXYS Number:
 778118
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More