• USA Home
  • Search Results

  • aobis23carboxy1oxopropylaminoethylpolyethyleneglycol123457455841111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α,ω-Bis{2-[(3-carboxy-1-oxopropyl)amino]ethyl}polyethylene glycol

3 Products
Synonym:   O,O′-Bis[2-(succinylamino)ethyl]polyethylene glycol
Image of <I>α,ω</I>-Bis{2-[(3-carboxy-1-oxopropyl)amino]ethyl}polyethylene glycol
CAS Number:
Linear Formula:
 HOOCCH2CH2CONH(CH2CH2O)nCH2CH2NHCOCH2CH2COOH
 MDL number:
 NACRES
 NA.23
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More