• USA Home
  • Search Results

  • apigenin7glucuronide446362974109111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Apigenin 7-glucuronide

2 Products
Synonym:   4′,5,7-Trihydroxyflavone 7-glucuronide, Apigenin 7-O-β-glucuronide, Scutellarin
Image of Apigenin 7-glucuronide
CAS Number:
Empirical Formula (Hill Notation):
 C21H18O11
Molecular Weight:
 446.36
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More