• USA Home
  • Search Results

  • apigenin7oglucuronide446362974109111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Apigenin 7-O-glucuronide

1 Products
Synonym:   4′,5,7-Trihydroxyflavone 7-glucuronide, Apigenin 7-glucuronide, Scutellarin
Image of Apigenin 7-<I>O</I>-glucuronide
CAS Number:
Empirical Formula (Hill Notation):
 C21H18O11
Molecular Weight:
 446.36
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More