• USA Home
  • Search Results

  • aprotininfrombovinelung651144908770111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aprotinin from bovine lung

9 Products
Synonym:   BPTI, Bovine pancreatic trypsin inhibitor, Trasylol, Trypsin inhibitor (basic)
Image of Aprotinin from bovine lung
CAS Number:
Empirical Formula (Hill Notation):
 C284H432N84O79S7
Molecular Weight:
 6511.44
 EC Number:
 MDL number:
 NACRES:
 NA.77
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Product Line
  Ice Shipping
  Application(s)
  Assay
  Solubility
  Biological Source
Show More

Product #
Description
A1153 Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid,
A6279 Aprotinin from bovine lung, saline solution, 3-7 TIU/mg protein,
A3428 Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid, BioReagent, suitable for cell culture,
10820 Aprotinin from bovine lung, lyophilized, ~80% (HPCE), crystalline (fine), white, ≥3500 U/mg,
A4529 Aprotinin from bovine lung, lyophilized powder, 3-7 TIU/mg solid,
A3886 Aprotinin from bovine lung,
A6106 Aprotinin from bovine lung, BioUltra, 3-8 TIU/mg solid, ≥98% (SDS-PAGE),
SRE0050 Aprotinin from bovine lung, saline solution, 3-7 TIU/mg protein, Suitable for manufacturing of diagnostic kits and reagents,
SRE0078 Aprotinin from bovine lung, lyophilized powder, ≥3 TIU/mg solid, Suitable for manufacturing of diagnostic kits and reagents,