• USA Home
  • Search Results

  • arachidicacid3125350630911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Arachidic acid

3 Products
Synonym:   Arachic acid, Arachidinic acid, Eicosanoic acid, Icosanoic acid, NSC 93983
Image of Arachidic acid
CAS Number:
Linear Formula:
 CH3(CH2)18COOH
Molecular Weight:
 312.53
 Beilstein/REAXYS Number:
 1788211
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Functional group
  Physical Form
  Special Grade
  Application(s)
  Assay
Show More