• USA Home
  • Search Results

  • arachidonylethanolamide347539442168811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Arachidonylethanolamide

1 Products
Synonym:   AEA, Anandamide (20:4, n-6), Anandamide, Arachidonic acid N-(hydroxyethyl)amide
Image of Arachidonylethanolamide
CAS Number:
Empirical Formula (Hill Notation):
 C22H37NO2
Molecular Weight:
 347.53
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More