• USA Home
  • Search Results

  • aripiprazolerelatedcompoundb2352888944320311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aripiprazole Related Compound B

2 Products
Synonym:   7-(4-Hydroxybutoxy)-3,4-dihydroquinolin-2(1H)-one
CAS Number:
Empirical Formula (Hill Notation):
 C13H17NO3
Molecular Weight:
 235.28
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More