• USA Home
  • Search Results

  • arseniciiioxide19784132753311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Arsenic(III) oxide

4 Products
Synonym:   Arsenic trioxide, Arsenous acid
Image of Arsenic(III) oxide
CAS Number:
Empirical Formula (Hill Notation):
 As2O3
Molecular Weight:
 197.84
 EC Number:
 MDL number:
 NACRES:
 NA.55
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Reaction Suitability
  Special Grade
  Vapor pressure
  Physical Form
Show More