• USA Home
  • Search Results

  • artemether298377196377411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Artemether

2 Products
Synonym:   Dihydroartemisinin methyl ether, Dihydroqinghaosu methyl ether, SM-224
Image of Artemether
CAS Number:
Empirical Formula (Hill Notation):
 C16H26O5
Molecular Weight:
 298.37
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More