• USA Home
  • Search Results

  • artemisinin282336396864911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Artemisinin

3 Products
Synonym:   Arteannuin, Qinghaosu
Image of Artemisinin
CAS Number:
Empirical Formula (Hill Notation):
 C15H22O5
Molecular Weight:
 282.33
 MDL number:
 NACRES:
 NA.79
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Activity Spectrum
  Titertest
Show More