• USA Home
  • Search Results

  • articainerelatedcompoundb2703511417652211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Articaine Related Compound B

2 Products
Synonym:   2-Carboxyarticaine, 4-Methyl-3-[2-(propylamino)propionamido]-2-thiophenecarboxylic acid, Articainic acid
CAS Number:
Empirical Formula (Hill Notation):
 C12H18N2O3S
Molecular Weight:
 270.35
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More