• USA Home
  • Search Results

  • ascorbicacid6palmitate4145313766611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ascorbic acid 6-palmitate

1 Products
Synonym:   6-O-Palmitoyl-L-ascorbic acid, L-Ascorbic acid 6-hexadecanoate, L-Ascorbyl palmitate, Ascorbic acid 6-palmitate, NSC 402451
Image of Ascorbic acid 6-palmitate
CAS Number:
Empirical Formula (Hill Notation):
 C22H38O7
Molecular Weight:
 414.53
 Beilstein/REAXYS Number:
 96552
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More