• USA Home
  • Search Results

  • ascorbylpalmitate4145313766611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ascorbyl palmitate

3 Products
Synonym:   6-O-Palmitoyl-L-ascorbic acid, L-Ascorbic acid 6-hexadecanoate, L-Ascorbyl palmitate, Ascorbic acid 6-palmitate, NSC 402451
Image of Ascorbyl palmitate
CAS Number:
Empirical Formula (Hill Notation):
 C22H38O7
Molecular Weight:
 414.53
 Beilstein/REAXYS Number:
 96552
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Application(s)
  Manufacturer designation
Show More