• USA Home
  • Search Results

  • asialogangliosidegm1frombovinebrain123457101219611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Asialoganglioside GM1 from bovine brain

1 Products
Synonym:   Ceramide tetrahexoside, Ganglioside GM1, asialo, Gangliotetraosyl ceramide
CAS Number:
 MDL number:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More