• USA Home
  • Search Results

  • aspirin180165078211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Aspirin

3 Products
Synonym:   2-Acetoxybenzoic acid, O-Acetylsalicylic acid, ASA, Acetylsalicylic acid, Aspirin
Image of Aspirin
CAS Number:
Linear Formula:
 2-(CH3CO2)C6H4CO2H
Molecular Weight:
 180.16
 Beilstein/REAXYS Number:
 779271
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More