• USA Home
  • Search Results

  • astaxanthin5968447261711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Astaxanthin

1 Products
Synonym:   (3S,3′S)-3,3′-Dihydroxy-β,β-Carotene-4,4′-dione, 3,3′-Dihydroxy-β-carotene-4,4′-dione, trans-Astaxanthin
CAS Number:
Empirical Formula (Hill Notation):
 C40H52O4
Molecular Weight:
 596.84
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More