• USA Home
  • Search Results

  • asulam23024333771111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Asulam

1 Products
Synonym:   Methyl N-(4-aminophenylsulfonyl)carbamate
Image of Asulam
CAS Number:
Empirical Formula (Hill Notation):
 C8H10N2O4S
Molecular Weight:
 230.24
 Beilstein/REAXYS Number:
 2697523
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More