• USA Home
  • Search Results

  • atenolol266342912268711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Atenolol

5 Products
Synonym:   (±)-4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetamide, 4-[2′-Hydroxy-3′-(isopropylamino)propoxy]phenylacetamide
Image of Atenolol
CAS Number:
Empirical Formula (Hill Notation):
 C14H22N2O3
Molecular Weight:
 266.34
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Titertest
  Assay
  Anal. Standard Form
  Special Grade
  Solubility
Show More