• USA Home
  • Search Results

  • aterpineol154259855511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

α-Terpineol

5 Products
Synonym:   alpha-Terpineol
Image of α-Terpineol
CAS Number:
Empirical Formula (Hill Notation):
 C10H18O
Molecular Weight:
 154.25
 Beilstein/REAXYS Number:
 2325137
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More