• USA Home
  • Search Results

  • atocopherol430711019141011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-α-Tocopherol

6 Products
Synonym:   DL-all-rac-α-Tocopherol, Vitamin E
Image of (±)-α-Tocopherol
CAS Number:
Empirical Formula (Hill Notation):
 C29H50O2
Molecular Weight:
 430.71
 Beilstein/REAXYS Number:
 94012
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
 eCl@ss:
 34058016
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More