• USA Home
  • Search Results

  • atraton21126161017911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Atraton

1 Products
Synonym:   Atrazine-methoxy
Image of Atraton
CAS Number:
Empirical Formula (Hill Notation):
 C9H17N5O
Molecular Weight:
 211.26
 Beilstein/REAXYS Number:
 613098
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More