• USA Home
  • Search Results

  • atrazine21568191224911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Atrazine

3 Products
Synonym:   2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine
Image of Atrazine
CAS Number:
Empirical Formula (Hill Notation):
 C8H14ClN5
Molecular Weight:
 215.68
 Beilstein/REAXYS Number:
 612020
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Package quantity
  Anal. Standard Form
  Physical Form
  Special Grade
Show More