• USA Home
  • Search Results

  • atrazine2hydroxy19724216368011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Atrazine-2-hydroxy

1 Products
Synonym:   2-Hydroxyatrazine
Image of Atrazine-2-hydroxy
CAS Number:
Empirical Formula (Hill Notation):
 C8H15N5O
Molecular Weight:
 197.24
 Beilstein/REAXYS Number:
 5870987
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Special Grade
Show More