• USA Home
  • Search Results

  • atrazinedesethyl18763619065411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Atrazine-desethyl

1 Products
Synonym:   2-Amino-4-chloro-6-isopropylamino-1,3,5-triazine, Desethylatrazine
Image of Atrazine-desethyl
CAS Number:
Empirical Formula (Hill Notation):
 C6H10ClN5
Molecular Weight:
 187.63
 Beilstein/REAXYS Number:
 610488
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More