• USA Home
  • Search Results

  • atrazinedesethyldesisopropyl14555339762411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Atrazine-desethyl-desisopropyl

1 Products
Synonym:   6-Chloro-2,4-diamino-1,3,5-triazine
Image of Atrazine-<WBR>desethyl-<WBR>desisopropyl
CAS Number:
Empirical Formula (Hill Notation):
 C3H4ClN5
Molecular Weight:
 145.55
 Beilstein/REAXYS Number:
 124340
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More