• USA Home
  • Search Results

  • atrazinedesethyldesisopropyl2hydroxy1271064592111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Atrazine-desethyl-desisopropyl-2-hydroxy

1 Products
Synonym:   4,6-Diamino-2-hydroxy-1,3,5-triazine, Ammeline
Image of Atrazine-<WBR>desethyl-<WBR>desisopropyl-<WBR>2-<WBR>hydroxy
CAS Number:
Empirical Formula (Hill Notation):
 C3H5N5O
Molecular Weight:
 127.10
 Beilstein/REAXYS Number:
 607753
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More