• USA Home
  • Search Results

  • atrazinedesisopropyl17360100728911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Atrazine-desisopropyl

1 Products
Synonym:   2-Amino-4-chloro-6-ethylamino-1,3,5-triazine
Image of Atrazine-<WBR>desisopropyl
CAS Number:
Empirical Formula (Hill Notation):
 C5H8ClN5
Molecular Weight:
 173.60
 Beilstein/REAXYS Number:
 8150
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More