• USA Home
  • Search Results

  • atropine289375155811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Atropine

4 Products
Synonym:   Hyoscyamine, Tropine tropate, endo-(±)-α-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester
Image of Atropine
CAS Number:
Empirical Formula (Hill Notation):
 C17H23NO3
Molecular Weight:
 289.37
 Beilstein/REAXYS Number:
 91260
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Anal. Standard Form
  Application(s)
  Manufacturer designation
  Physical Form
  Special Grade
Show More