• USA Home
  • Search Results

  • atropinesulfate69483590899611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Atropine sulfate

4 Products
Synonym:   α-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, Atropine sulfate salt monohydrate, Tropine tropate
Image of Atropine sulfate
CAS Number:
Linear Formula:
 (C17H23NO3)2 · H2SO4 · H2O
Molecular Weight:
 694.83
 Beilstein/REAXYS Number:
 6109275
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Anal. Standard Form
  Physical Form
  Manufacturer designation
Show More