• USA Home
  • Search Results

  • atropinesulfatesaltmonohydrate69483590899611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Atropine sulfate salt monohydrate

1 Products
Synonym:   α-(Hydroxymethyl)benzeneacetic acid 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, Tropine tropate
Image of Atropine sulfate salt monohydrate
CAS Number:
Linear Formula:
 (C17H23NO3)2 · H2SO4 · H2O
Molecular Weight:
 694.83
 Beilstein/REAXYS Number:
 6109275
 EC Number:
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More