• USA Home
  • Search Results

  • avicularin4343557230511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Avicularin

2 Products
Synonym:   3,3′,4′,5,7-Pentahydroxyflavone 3-O-α-L-arabinofuranoside, Avicularoside, Quercetin 3-O-α-L-arabinofuranoside
Image of Avicularin
CAS Number:
Empirical Formula (Hill Notation):
 C20H18O11
Molecular Weight:
 434.35
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Application(s)
Show More