• USA Home
  • Search Results

  • azaconazole300146020731011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Azaconazole

1 Products
Synonym:   1-[2-(2,4-Dichlorophenyl)-1,3-dioxolan-2-ylmethyl]-1H-1,2,4-triazole
Image of Azaconazole
CAS Number:
Empirical Formula (Hill Notation):
 C12H11Cl2N3O2
Molecular Weight:
 300.14
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More