• USA Home
  • Search Results

  • azadirachtin720711114117611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Azadirachtin

2 Products
Image of Azadirachtin
CAS Number:
Empirical Formula (Hill Notation):
 C35H44O16
Molecular Weight:
 720.71
 MDL number:
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Manufacturer designation
Show More