• USA Home
  • Search Results

  • azathioprinerelatedcompounda14212453154811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Azathioprine Related Compound A

1 Products
Synonym:   1-Methyl-4-nitro-1H-imidazol-5-amine
CAS Number:
Empirical Formula (Hill Notation):
 C4H6N4O2
Molecular Weight:
 142.12
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More