• USA Home
  • Search Results

  • azelaicacid1882212399911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Azelaic acid

6 Products
Synonym:   Nonanedioic acid
Image of Azelaic acid
CAS Number:
Linear Formula:
 HO2C(CH2)7CO2H
Molecular Weight:
 188.22
 Beilstein/REAXYS Number:
 1101094
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Vapor pressure
  Transition temperature
  Application(s)
  Special Grade
  Melting Point
  Potency
Show More