• USA Home
  • Search Results

  • azetidinehydrochloride93563652039511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Azetidine hydrochloride

1 Products
Synonym:   Trimethyleneimine hydrochloride
Image of Azetidine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C3H7N · HCl
Molecular Weight:
 93.56
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More