• USA Home
  • Search Results

  • azinphosmethyl317328650011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Azinphos-methyl

1 Products
Synonym:   Guthion®, Methyltriazotin
Image of Azinphos-methyl
CAS Number:
Empirical Formula (Hill Notation):
 C10H12N3O3PS2
Molecular Weight:
 317.32
 Beilstein/REAXYS Number:
 280476
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More